Product Details

SNP ID
rs142231846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:25062236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTTCTGTTCTGTGAGAACATC[C/T]CTTCATGGTTTGCAAGAATCTTCAG
Phenotype
MIM: 610974
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF521 PubMed Links

Gene Details

Gene
ZNF521
Gene Name
zinc finger protein 521
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308225.1 4622 UTR 3 NP_001295154.1
NM_015461.2 4622 UTR 3 NP_056276.1
XM_011525909.2 4622 UTR 3 XP_011524211.1
XM_011525910.2 4622 Intron XP_011524212.1
XM_011525911.1 4622 UTR 3 XP_011524213.1
XM_017025697.1 4622 UTR 3 XP_016881186.1
XM_017025698.1 4622 UTR 3 XP_016881187.1

View Full Product Details