Product Details

SNP ID
rs142943775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:62044410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAAAGTTCATGATTCTCATTCC[A/C]TCAACATCACTGTTTGCTACCATAG
Phenotype
MIM: 606097
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PIGN PubMed Links

Gene Details

Gene
PIGN
Gene Name
phosphatidylinositol glycan anchor biosynthesis class N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012327.5 3846 UTR 3 NP_036459.1
NM_176787.4 3846 UTR 3 NP_789744.1
XM_011525889.1 3846 UTR 3 XP_011524191.1
XM_011525890.1 3846 UTR 3 XP_011524192.1
XM_011525891.1 3846 UTR 3 XP_011524193.1
XM_011525892.1 3846 UTR 3 XP_011524194.1
XM_011525893.1 3846 UTR 3 XP_011524195.1
XM_011525894.1 3846 UTR 3 XP_011524196.1
XM_011525895.1 3846 UTR 3 XP_011524197.1
XM_011525896.1 3846 UTR 3 XP_011524198.1
XM_011525898.1 3846 Intron XP_011524200.1
XM_017025685.1 3846 UTR 3 XP_016881174.1
XM_017025686.1 3846 UTR 3 XP_016881175.1

View Full Product Details