Product Details
- SNP ID
-
rs143026087
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:22181524 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAACTACCACCTTATGGCGCAGAAA[C/T]GCCGAGGGTGAACCCGTGTGCAATG
- Phenotype
-
MIM: 601656
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GATA6
PubMed Links
Gene Details
- Gene
- GATA6
- Gene Name
- GATA binding protein 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005257.5 |
1657 |
Silent Mutation |
AAC,AAT |
N458N |
NP_005248.2 |
View Full Product Details