Product Details
- SNP ID
-
rs143090156
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:43273644 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTCCTTCAGATAATTCAATTCCC[A/G]AGAGAGGAATTAGAACTTCCCCAAT
- Phenotype
-
MIM: 600103
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SYT4
PubMed Links
Gene Details
- Gene
- SYT4
- Gene Name
- synaptotagmin 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020783.3 |
1154 |
Missense Mutation |
|
|
NP_065834.1 |
View Full Product Details