Product Details

SNP ID
rs143447794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63588727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTGATCTTTTCAAAGAGCTGAA[A/G]AAAACAAATGATGGCAACATCTTCT
Phenotype
MIM: 604445
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINB13 PubMed Links

Gene Details

Gene
SERPINB13
Gene Name
serpin family B member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307923.1 228 Silent Mutation AAA,AAG K20K NP_001294852.1
NM_012397.3 228 Silent Mutation AAA,AAG K20K NP_036529.1
XM_005266707.3 228 UTR 5 XP_005266764.1
XM_011526029.2 228 Silent Mutation AAA,AAG K20K XP_011524331.1

View Full Product Details