Product Details
- SNP ID
-
rs143817151
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:13826365 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATGTTCTTCGCTATGCTGTTCCT[C/T]CTGGTGTCTCTGTACATACACATGT
- Phenotype
-
MIM: 600042
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MC5R
PubMed Links
Gene Details
- Gene
- MC5R
- Gene Name
- melanocortin 5 receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005913.2 |
822 |
Silent Mutation |
CTC,CTT |
L200L |
NP_005904.1 |
View Full Product Details