Product Details

SNP ID
rs144221180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9141042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGCTCTGCCTCTAATTAAGAGTG[C/T]TGGTTTTCGAAGAGTTTGAAGGCGG
Phenotype
MIM: 610616 MIM: 600532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD12 PubMed Links

Gene Details

Gene
ANKRD12
Gene Name
ankyrin repeat domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083625.2 Intron NP_001077094.1
NM_001204056.1 Intron NP_001190985.1
NM_015208.4 Intron NP_056023.3
XM_005258092.3 Intron XP_005258149.1
XM_005258093.3 Intron XP_005258150.1
XM_005258095.2 Intron XP_005258152.1
XM_005258096.2 Intron XP_005258153.1
XM_011525638.2 Intron XP_011523940.1
XM_011525639.2 Intron XP_011523941.1
XM_017025661.1 Intron XP_016881150.1
XM_017025662.1 Intron XP_016881151.1
XM_017025663.1 Intron XP_016881152.1
XM_017025664.1 Intron XP_016881153.1
XM_017025665.1 Intron XP_016881154.1
XM_017025666.1 Intron XP_016881155.1
XM_017025667.1 Intron XP_016881156.1
XM_017025668.1 Intron XP_016881157.1
XM_017025669.1 Intron XP_016881158.1
XM_017025670.1 Intron XP_016881159.1
Gene
NDUFV2
Gene Name
NADH:ubiquinone oxidoreductase core subunit V2
There are no transcripts associated with this gene.

Gene
NDUFV2-AS1
Gene Name
NDUFV2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details