Product Details

SNP ID
rs144795065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:59328262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGTACAACTGTCTGATATAAA[C/T]TGTTACCATAATCACAATCAGGAAG
Phenotype
MIM: 609586 MIM: 601567
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPLX4 PubMed Links

Gene Details

Gene
CPLX4
Gene Name
complexin 4
There are no transcripts associated with this gene.

Gene
LMAN1
Gene Name
lectin, mannose binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005570.3 4396 UTR 3 NP_005561.1

View Full Product Details