Product Details

SNP ID
rs145110091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:41963444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGCTCAAAAAATCTCATTTTTG[G/T]TCAGTAGGAGACCCTTCAAGTAGAC
Phenotype
MIM: 602609
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PIK3C3 PubMed Links

Gene Details

Gene
PIK3C3
Gene Name
phosphatidylinositol 3-kinase catalytic subunit type 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308020.1 Intron NP_001294949.1
NM_002647.3 Intron NP_002638.2
XM_011526031.2 Intron XP_011524333.1

View Full Product Details