Product Details

SNP ID
rs145235987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54360630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCAGGAATCTTCAGTATCTGCC[C/T]TGCTGGCAAGCTGCACCCTGAGTGG
Phenotype
MIM: 613258 MIM: 607051
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C18orf54 PubMed Links

Gene Details

Gene
C18orf54
Gene Name
chromosome 18 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288980.1 192 Silent Mutation CTG,TTG L20L NP_001275909.1
NM_001288981.1 192 Silent Mutation CTG,TTG L20L NP_001275910.1
NM_001288982.1 192 Silent Mutation CTG,TTG L20L NP_001275911.1
NM_173529.5 192 Silent Mutation CTG,TTG L20L NP_775800.3
XM_005258201.1 192 Silent Mutation CTG,TTG L20L XP_005258258.1
XM_005258203.1 192 Silent Mutation CTG,TTG L20L XP_005258260.1
XM_017025567.1 192 Silent Mutation CTG,TTG L20L XP_016881056.1
Gene
STARD6
Gene Name
StAR related lipid transfer domain containing 6
There are no transcripts associated with this gene.

View Full Product Details