Product Details

SNP ID
rs145453378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50270661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGAATCCTATGCAGCACCTGGG[A/G]TCTGCAGGAAACCCAGTTTCTGTAT
Phenotype
MIM: 614759 MIM: 156535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP53 PubMed Links

Gene Details

Gene
CFAP53
Gene Name
cilia and flagella associated protein 53
There are no transcripts associated with this gene.

Gene
MBD1
Gene Name
methyl-CpG binding domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204136.1 2779 Intron NP_001191065.1
NM_001204137.1 2779 Intron NP_001191066.1
NM_001204138.1 2779 Intron NP_001191067.1
NM_001204139.1 2779 Intron NP_001191068.1
NM_001204140.1 2779 Intron NP_001191069.1
NM_001204141.1 2779 Intron NP_001191070.1
NM_001204142.1 2779 Intron NP_001191071.1
NM_001204143.1 2779 Intron NP_001191072.1
NM_001204151.2 2779 Intron NP_001191080.1
NM_001323942.1 2779 Intron NP_001310871.1
NM_001323947.1 2779 Intron NP_001310876.1
NM_001323949.1 2779 Intron NP_001310878.1
NM_001323950.1 2779 Intron NP_001310879.1
NM_001323951.1 2779 Intron NP_001310880.1
NM_001323952.1 2779 Intron NP_001310881.1
NM_001323953.1 2779 Intron NP_001310882.1
NM_001323954.1 2779 Intron NP_001310883.1
NM_002384.2 2779 Intron NP_002375.1
NM_015844.2 2779 Intron NP_056669.2
NM_015845.3 2779 Intron NP_056670.2
NM_015846.3 2779 Intron NP_056671.2
NM_015847.3 2779 Intron NP_056723.2
XM_005258271.2 2779 UTR 3 XP_005258328.1
XM_006722456.2 2779 Intron XP_006722519.1
XM_011525991.1 2779 Intron XP_011524293.1
XM_011525993.2 2779 Intron XP_011524295.1
XM_011525994.2 2779 Intron XP_011524296.1
XM_011525998.2 2779 Intron XP_011524300.1
XM_011525999.1 2779 Intron XP_011524301.1
XM_011526001.1 2779 Intron XP_011524303.1
XM_011526002.1 2779 UTR 3 XP_011524304.1
XM_011526003.2 2779 Intron XP_011524305.1
XM_011526006.1 2779 Intron XP_011524308.1
XM_011526007.1 2779 UTR 3 XP_011524309.1
XM_017025751.1 2779 Intron XP_016881240.1
XM_017025752.1 2779 Intron XP_016881241.1
XM_017025753.1 2779 Intron XP_016881242.1
XM_017025754.1 2779 Intron XP_016881243.1
XM_017025755.1 2779 Intron XP_016881244.1
XM_017025756.1 2779 Intron XP_016881245.1
XM_017025757.1 2779 UTR 3 XP_016881246.1
XM_017025758.1 2779 Intron XP_016881247.1
XM_017025759.1 2779 Intron XP_016881248.1
XM_017025760.1 2779 Intron XP_016881249.1
XM_017025761.1 2779 Intron XP_016881250.1
XM_017025762.1 2779 Intron XP_016881251.1
XM_017025763.1 2779 UTR 3 XP_016881252.1
XM_017025764.1 2779 Intron XP_016881253.1
XM_017025765.1 2779 Intron XP_016881254.1
XM_017025766.1 2779 Intron XP_016881255.1
XM_017025767.1 2779 Intron XP_016881256.1
XM_017025768.1 2779 Intron XP_016881257.1
XM_017025769.1 2779 Intron XP_016881258.1
XM_017025770.1 2779 UTR 3 XP_016881259.1
XM_017025771.1 2779 Intron XP_016881260.1
XM_017025772.1 2779 Intron XP_016881261.1
XM_017025773.1 2779 UTR 3 XP_016881262.1
XM_017025774.1 2779 Intron XP_016881263.1
XM_017025775.1 2779 Intron XP_016881264.1
XM_017025776.1 2779 UTR 3 XP_016881265.1
XM_017025777.1 2779 Intron XP_016881266.1

View Full Product Details