Product Details

SNP ID
rs145745512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:41957570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATTGTTGGTCTTGTGCTTTCAG[A/G]GGAAGCTTGGAAGGGAAGAGAGAAC
Phenotype
MIM: 602609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIK3C3 PubMed Links

Gene Details

Gene
PIK3C3
Gene Name
phosphatidylinositol 3-kinase catalytic subunit type 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308020.1 163 Intron NP_001294949.1
NM_002647.3 163 Missense Mutation ATA,ATG I23M NP_002638.2
XM_011526031.2 163 UTR 5 XP_011524333.1

View Full Product Details