Product Details

SNP ID
rs145852832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:32680452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGTGCACTGCTCCCGCATGAGC[C/T]GCCAGAGGCTGTGGCAAAGAGGACA
Phenotype
MIM: 613772
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL14 PubMed Links

Gene Details

Gene
KLHL14
Gene Name
kelch like family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020805.2 1774 Silent Mutation GCA,GCG A462A NP_065856.1
XM_011526109.2 1774 Intron XP_011524411.2

View Full Product Details