Product Details

SNP ID
rs146231086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:10527818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAAGAGTTGGGGCCAGACTGTG[A/C]AGGGACAGCTTCTCACATGGCTATT
Phenotype
MIM: 603216
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NAPG PubMed Links

Gene Details

Gene
NAPG
Gene Name
NSF attachment protein gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003826.2 Intron NP_003817.1
XM_011525754.2 Intron XP_011524056.1
XM_011525756.2 Intron XP_011524058.1
XM_017026063.1 Intron XP_016881552.1

View Full Product Details