Product Details

SNP ID
rs146480648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35242612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGTACTCAATCCTGCCAAGAA[C/T]TGTTTCGTCAGCAATTCAGAAAATT
Phenotype
MIM: 609601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF397 PubMed Links

Gene Details

Gene
ZNF397
Gene Name
zinc finger protein 397
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135178.2 298 Silent Mutation CTG,TTG L48L NP_001128650.1
NM_032347.2 298 Silent Mutation CTG,TTG L48L NP_115723.1
XM_006722558.2 298 Silent Mutation CTG,TTG L48L XP_006722621.1
XM_011526229.2 298 Silent Mutation CTG,TTG L48L XP_011524531.1
XM_011526230.2 298 Silent Mutation CTG,TTG L48L XP_011524532.1
XM_011526231.2 298 Silent Mutation CTG,TTG L48L XP_011524533.1
XM_017026042.1 298 Silent Mutation CTG,TTG L48L XP_016881531.1
Gene
ZSCAN30
Gene Name
zinc finger and SCAN domain containing 30
There are no transcripts associated with this gene.

View Full Product Details