Product Details

SNP ID
rs148323869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:23463003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCAAAATACAATATCTTGTTCT[C/T]TGGCTGATGTAATGAGTGAACAGCT
Phenotype
MIM: 603579
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RIOK3 PubMed Links

Gene Details

Gene
RIOK3
Gene Name
RIO kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003831.3 720 Silent Mutation CTG,TTG L35L NP_003822.2
XM_011526242.1 720 UTR 5 XP_011524544.1
XM_011526243.1 720 UTR 5 XP_011524545.1

View Full Product Details