Product Details

SNP ID
rs149877496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:23703387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGAGTCTACATAATTCAACTAA[C/T]GTTCCATAGCTGTGACTGGTAGCCA
Phenotype
MIM: 600805
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAMA3 PubMed Links

Gene Details

Gene
LAMA3
Gene Name
laminin subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000227.4 Intron NP_000218.3
NM_001127717.2 Intron NP_001121189.2
NM_001127718.2 Intron NP_001121190.2
NM_001302996.1 Intron NP_001289925.1
NM_198129.2 Intron NP_937762.2
XM_011525978.2 Intron XP_011524280.1
XM_011525979.2 Intron XP_011524281.1
XM_011525980.2 Intron XP_011524282.1
XM_011525981.2 Intron XP_011524283.1
XM_011525982.2 Intron XP_011524284.1
XM_017025743.1 Intron XP_016881232.1
XM_017025744.1 Intron XP_016881233.1

View Full Product Details