Product Details

SNP ID
rs149991068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:44688406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTAGCAGACTAATGAAATCTGC[A/G]TGGCAAGTCCTGTACAGTTGTTGGT
Phenotype
MIM: 611060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927943 PubMed Links

Gene Details

Gene
LOC101927943
Gene Name
uncharacterized LOC101927943
There are no transcripts associated with this gene.

Gene
SETBP1
Gene Name
SET binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130110.1 Intron NP_001123582.1
NM_015559.2 Intron NP_056374.2

View Full Product Details