Product Details

SNP ID
rs150347076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63891518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCATCTGGCAAAAGCAAGCCCCA[C/T]CCAGAACCTCTTCCTCTCCCCATGG
Phenotype
MIM: 173390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINB2 PubMed Links

Gene Details

Gene
SERPINB2
Gene Name
serpin family B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143818.1 149 Missense Mutation ACC,ATC T25I NP_001137290.1
NM_002575.2 149 Missense Mutation ACC,ATC T25I NP_002566.1

View Full Product Details