Product Details

SNP ID
rs150798338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:58482001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTGTAGTGCTTTAAACTGATCA[A/G]TGAAGGTCATGGAACAGTTGCCTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALPK2 PubMed Links

Gene Details

Gene
ALPK2
Gene Name
alpha kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052947.3 5667 Missense Mutation ACT,ATT T2112I NP_443179.3
XM_011525801.2 5667 UTR 3 XP_011524103.1
XM_011525802.2 5667 Missense Mutation ACT,ATT T2018I XP_011524104.1
XM_017025535.1 5667 Intron XP_016881024.1
XM_017025536.1 5667 Intron XP_016881025.1
XM_017025537.1 5667 Intron XP_016881026.1
XM_017025538.1 5667 Missense Mutation ACT,ATT T1443I XP_016881027.1
XM_017025539.1 5667 Missense Mutation ACT,ATT T1443I XP_016881028.1

View Full Product Details