Product Details
- SNP ID
-
hCV165219713
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:53408057 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTGATAATCAAGTTGTGAAGTCT[G/A]TCCACGATGCTTCCTTGGTTTCAAC
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF765
PubMed Links
Gene Details
- Gene
- ZNF765
- Gene Name
- zinc finger protein 765
There are no transcripts associated with this gene.
View Full Product Details