Product Details

SNP ID
rs137886346
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53068811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTTGCAAGTTGTGATGTTTGAG[C/T]GAAGACTTTACCACATTCATTACAC
Phenotype
MIM: 600398
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZNF160 PubMed Links
Additional Information
For this assay, SNP(s) [rs78382850] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ZNF160
Gene Name
zinc finger protein 160
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102603.1 2135 Missense Mutation NP_001096073.1
NM_001322125.1 2135 Intron NP_001309054.1
NM_001322126.1 2135 Intron NP_001309055.1
NM_001322128.1 2135 Missense Mutation NP_001309057.1
NM_001322129.1 2135 Missense Mutation NP_001309058.1
NM_001322130.1 2135 Missense Mutation NP_001309059.1
NM_001322131.1 2135 Missense Mutation NP_001309060.1
NM_001322132.1 2135 Missense Mutation NP_001309061.1
NM_001322133.1 2135 Missense Mutation NP_001309062.1
NM_001322134.1 2135 Missense Mutation NP_001309063.1
NM_001322135.1 2135 Missense Mutation NP_001309064.1
NM_001322136.1 2135 Missense Mutation NP_001309065.1
NM_001322137.1 2135 Missense Mutation NP_001309066.1
NM_001322138.1 2135 Missense Mutation NP_001309067.1
NM_001322139.1 2135 Missense Mutation NP_001309068.1
NM_033288.3 2135 Missense Mutation NP_150630.2
NM_198893.2 2135 Missense Mutation NP_942596.1
XM_017027445.1 2135 Missense Mutation XP_016882934.1
XM_017027446.1 2135 Missense Mutation XP_016882935.1
XM_017027447.1 2135 Intron XP_016882936.1
XM_017027448.1 2135 Intron XP_016882937.1

View Full Product Details