Product Details

SNP ID
rs137968069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57941097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTGCATTCATAAGGCCTTTCTC[C/T]GGTATGAACTCTTCGGTGTTTAACG
Phenotype
MIM: 606956
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF256 PubMed Links

Gene Details

Gene
ZNF256
Gene Name
zinc finger protein 256
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005773.2 1946 Missense Mutation NP_005764.2
XM_017026138.1 1946 Missense Mutation XP_016881627.1
Gene
ZNF418
Gene Name
zinc finger protein 418
There are no transcripts associated with this gene.

View Full Product Details