Product Details

SNP ID
rs137977632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10274812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTCCCCCTCAAAAGTCATCCTG[C/T]CCCGGGGAGGCTCCGTGCTGGTGAC
Phenotype
MIM: 147840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ICAM1 PubMed Links

Gene Details

Gene
ICAM1
Gene Name
intercellular adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000201.2 434 Missense Mutation CCC,TCC P39S NP_000192.2

View Full Product Details