Product Details

SNP ID
rs138158370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12075654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTAAAGAATGTGGAAAAACCTT[C/T]ATATCCCATTCAAGCATTCAAAGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF844 PubMed Links

Gene Details

Gene
ZNF844
Gene Name
zinc finger protein 844
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136501.2 555 Silent Mutation TTC,TTT F178F NP_001129973.1
XM_017026664.1 555 Silent Mutation TTC,TTT F180F XP_016882153.1

View Full Product Details