Product Details

SNP ID
rs138168830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38932418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGCCTGGGGCCCCCCAAGCGGCC[A/G]CCTCGGAAGCTGGGCCCCACGGAAG
Phenotype
MIM: 609094 MIM: 603021 MIM: 612804
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO17 PubMed Links

Gene Details

Gene
FBXO17
Gene Name
F-box protein 17
There are no transcripts associated with this gene.

Gene
MRPS12
Gene Name
mitochondrial ribosomal protein S12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021107.1 445 Silent Mutation CCA,CCG P45P NP_066930.1
NM_033362.3 445 Silent Mutation CCA,CCG P45P NP_203526.1
NM_033363.1 445 Silent Mutation CCA,CCG P45P NP_203527.1
Gene
SARS2
Gene Name
seryl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details