Product Details

SNP ID
rs138205709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4306026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTATCTACTCCCTGAAACAGAT[C/G]CTGCTGAACGTGGAGGTGAAGGCGG
Phenotype
MIM: 609828 MIM: 614715
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FSD1 PubMed Links

Gene Details

Gene
FSD1
Gene Name
fibronectin type III and SPRY domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024333.2 248 Missense Mutation ATC,ATG I32M NP_077309.1
XM_005259644.2 248 Missense Mutation ATC,ATG I32M XP_005259701.1
Gene
TMIGD2
Gene Name
transmembrane and immunoglobulin domain containing 2
There are no transcripts associated with this gene.

View Full Product Details