Product Details

SNP ID
rs138256115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49559993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAACAGCAGTCGAAGTCCTTCAC[A/G]GCATCCCGGCTCAGTCGAATGTTCT
Phenotype
MIM: 616759
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOSIP PubMed Links

Gene Details

Gene
NOSIP
Gene Name
nitric oxide synthase interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270960.1 252 Silent Mutation GCC,GCT A39A NP_001257889.1
NM_015953.4 252 Silent Mutation GCC,GCT A39A NP_057037.1
XM_005258964.4 252 Silent Mutation GCC,GCT A39A XP_005259021.1
XM_006723235.3 252 Silent Mutation GCC,GCT A39A XP_006723298.1
XM_011527015.2 252 Silent Mutation GCC,GCT A40A XP_011525317.1
XM_011527017.2 252 Silent Mutation GCC,GCT A40A XP_011525319.1
XM_017026851.1 252 Silent Mutation GCC,GCT A39A XP_016882340.1
XM_017026852.1 252 Silent Mutation GCC,GCT A39A XP_016882341.1

View Full Product Details