Product Details

SNP ID
rs138308187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11506244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCTTCTTCCTGGTGGTCCTCGGG[C/T]AGGGGCGGCTCCTCCAGCCCTGCAG
Phenotype
MIM: 608388 MIM: 611371
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ECSIT PubMed Links

Gene Details

Gene
ECSIT
Gene Name
ECSIT signalling integrator
There are no transcripts associated with this gene.

Gene
ZNF653
Gene Name
zinc finger protein 653
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138783.3 Intron NP_620138.2

View Full Product Details