Product Details

SNP ID
rs138391803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56222722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACCTGTTACGTCAATACTCTTG[C/T]GTAGCAGAAAGTCCTCTCCCTGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN5A PubMed Links

Gene Details

Gene
ZSCAN5A
Gene Name
zinc finger and SCAN domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322061.1 957 Missense Mutation NP_001308990.1
NM_001322062.1 957 Intron NP_001308991.1
NM_001322064.1 957 Missense Mutation NP_001308993.1
NM_001322065.1 957 Missense Mutation NP_001308994.1
NM_001322066.1 957 Missense Mutation NP_001308995.1
NM_001322067.1 957 Missense Mutation NP_001308996.1
NM_001322068.1 957 Missense Mutation NP_001308997.1
NM_001322069.1 957 Missense Mutation NP_001308998.1
NM_001322070.1 957 Missense Mutation NP_001308999.1
NM_001322072.1 957 Missense Mutation NP_001309001.1
NM_001322073.1 957 Missense Mutation NP_001309002.1
NM_001322074.1 957 Missense Mutation NP_001309003.1
NM_001322075.1 957 Missense Mutation NP_001309004.1
NM_001322076.1 957 Missense Mutation NP_001309005.1
NM_001322077.1 957 Missense Mutation NP_001309006.1
NM_001322078.1 957 Missense Mutation NP_001309007.1
NM_024303.2 957 Intron NP_077279.1
XM_005259254.1 957 Intron XP_005259311.1
XM_006723377.3 957 Missense Mutation XP_006723440.1
XM_011527308.1 957 Intron XP_011525610.1
XM_017027298.1 957 Missense Mutation XP_016882787.1
XM_017027299.1 957 Missense Mutation XP_016882788.1

View Full Product Details