Product Details

SNP ID
rs138452209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51411181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCTGGGTCTGACGCCTGGGAAG[C/T]TGAGCGTGGCATAATGGAGCTCCTC
Phenotype
MIM: 606091
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100129083 PubMed Links

Gene Details

Gene
LOC100129083
Gene Name
uncharacterized LOC100129083
There are no transcripts associated with this gene.

Gene
SIGLEC10
Gene Name
sialic acid binding Ig like lectin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171156.1 1775 Missense Mutation AAC,AGC N613S NP_001164627.1
NM_001171157.1 1775 Intron NP_001164628.1
NM_001171158.1 1775 Missense Mutation AAC,AGC N528S NP_001164629.1
NM_001171159.1 1775 Missense Mutation AAC,AGC N518S NP_001164630.1
NM_001171161.1 1775 Missense Mutation AAC,AGC N428S NP_001164632.1
NM_001322105.1 1775 Missense Mutation AAC,AGC N486S NP_001309034.1
NM_033130.4 1775 Missense Mutation AAC,AGC N671S NP_149121.2
XM_005259366.4 1775 Missense Mutation AAC,AGC N581S XP_005259423.1
XM_005259367.3 1775 Missense Mutation AAC,AGC N523S XP_005259424.1
XM_011527439.1 1775 Missense Mutation AAC,AGC N623S XP_011525741.1

View Full Product Details