Product Details

SNP ID
rs138510982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9214002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTTCAGCATGGGGGTGACCATG[G/T]CGTACATCACTGAGGCGGTGGAGCT
Phenotype
MIM: 611538
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR7D4 PubMed Links

Gene Details

Gene
OR7D4
Gene Name
olfactory receptor family 7 subfamily D member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005191.2 Intron NP_001005191.1

View Full Product Details