Product Details

SNP ID
rs138603798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53882542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGCGATTCAGAGGGGGGACCCCG[A/G]CCCCTGTTTTGCAGAAAGGGGGCCC
Phenotype
MIM: 609959 MIM: 176980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYADM PubMed Links

Gene Details

Gene
MYADM
Gene Name
myeloid associated differentiation marker
There are no transcripts associated with this gene.

Gene
PRKCG
Gene Name
protein kinase C gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316329.1 359 Silent Mutation CGA,CGG R16R NP_001303258.1
NM_002739.4 359 Silent Mutation CGA,CGG R16R NP_002730.1

View Full Product Details