Product Details

SNP ID
rs138722048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41354833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGTGTCCCCATGCAGCAGCTGC[A/G]GCCCACCACCCACGAAAGCCCGTGC
Phenotype
MIM: 611951 MIM: 190180
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B9D2 PubMed Links

Gene Details

Gene
B9D2
Gene Name
B9 protein domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030578.3 615 Missense Mutation CCG,CTG P132L NP_085055.2
XM_011527349.2 615 Missense Mutation CCG,CTG P132L XP_011525651.1
XM_011527350.1 615 Missense Mutation CCG,CTG P79L XP_011525652.1
Gene
TGFB1
Gene Name
transforming growth factor beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011527242.1 615 Intron XP_011525544.1
Gene
TMEM91
Gene Name
transmembrane protein 91
There are no transcripts associated with this gene.

View Full Product Details