Product Details

SNP ID
rs138730527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12737528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGGCGGCAGGGGTGGCCGGGT[G/T]GGGGGTTGAGGCAGAGGAGTTCGAA
Phenotype
MIM: 601913
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASNA1 PubMed Links

Gene Details

Gene
ASNA1
Gene Name
arsA arsenite transporter, ATP-binding, homolog 1 (bacterial)
There are no transcripts associated with this gene.

Gene
C19orf43
Gene Name
chromosome 19 open reading frame 43
There are no transcripts associated with this gene.

View Full Product Details