Product Details

SNP ID
rs138753275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50883365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTGGTTCCCACTACATCCACAT[C/T]CAGTTCACCTATTTGGCCCATGCTG
Phenotype
MIM: 147960
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLK2 PubMed Links

Gene Details

Gene
KLK2
Gene Name
kallikrein related peptidase 2
There are no transcripts associated with this gene.

Gene
KLKP1
Gene Name
kallikrein pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details