Product Details

SNP ID
rs138896705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6665066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGCTGCTGTCCCACCAGACCC[A/G]GGAGCTGCTGGTGGCCCGTCCGCAG
Phenotype
MIM: 604520
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFSF14 PubMed Links

Gene Details

Gene
TNFSF14
Gene Name
tumor necrosis factor superfamily member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003807.4 965 Missense Mutation CGG,TGG R195W NP_003798.2
NM_172014.3 965 Missense Mutation CGG,TGG R159W NP_742011.2
XM_017027417.1 965 Missense Mutation CGG,TGG R195W XP_016882906.1
XM_017027418.1 965 Intron XP_016882907.1

View Full Product Details