Product Details

SNP ID
rs138978355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5914411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCAGAGCCCAGACCAAGCATGG[A/G]CGCCGTGGATGCCACCATGGAGAAA
Phenotype
MIM: 114212 MIM: 603327
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPS PubMed Links

Gene Details

Gene
CAPS
Gene Name
calcyphosine
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004058.4 693 Missense Mutation GAC,GGC D88G NP_004049.2
NM_080590.3 693 Missense Mutation GAC,GGC D88G NP_542157.2
Gene
RANBP3
Gene Name
RAN binding protein 3
There are no transcripts associated with this gene.

Gene
VMAC
Gene Name
vimentin-type intermediate filament associated coiled-coil protein
There are no transcripts associated with this gene.

View Full Product Details