Product Details

SNP ID
rs139009034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1482419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTCACCTGGGGGCCTGTAGGCC[A/G]CGCTGTCATGTCCTCGGCCGTCCCA
Phenotype
MIM: 612034 MIM: 600487 MIM: 609346
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APC2 PubMed Links

Gene Details

Gene
APC2
Gene Name
APC2, WNT signaling pathway regulator
There are no transcripts associated with this gene.

Gene
C19orf25
Gene Name
chromosome 19 open reading frame 25
There are no transcripts associated with this gene.

Gene
PCSK4
Gene Name
proprotein convertase subtilisin/kexin type 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017573.4 1417 Silent Mutation NP_060043.2
XM_005259586.1 1417 Missense Mutation XP_005259643.1
XM_011528085.2 1417 Missense Mutation XP_011526387.1
XM_011528086.2 1417 Missense Mutation XP_011526388.1
XM_011528087.2 1417 Missense Mutation XP_011526389.1
XM_011528088.2 1417 Missense Mutation XP_011526390.1
XM_011528089.2 1417 Missense Mutation XP_011526391.1
XM_011528090.2 1417 Missense Mutation XP_011526392.1
XM_011528091.2 1417 Missense Mutation XP_011526393.1
XM_011528092.2 1417 Missense Mutation XP_011526394.1
XM_011528093.2 1417 Missense Mutation XP_011526395.1
XM_011528094.1 1417 Missense Mutation XP_011526396.1
XM_011528095.2 1417 UTR 3 XP_011526397.1
XM_011528096.1 1417 Missense Mutation XP_011526398.1
XM_017026897.1 1417 Missense Mutation XP_016882386.1
Gene
REEP6
Gene Name
receptor accessory protein 6
There are no transcripts associated with this gene.

View Full Product Details