Product Details

SNP ID
rs139018520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47340903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGCCTGGCCATCGACCCGCTGC[A/G]CGTGGCCCCGCTCCCACTGTATGCC
Phenotype
MIM: 609949 MIM: 615475
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C5AR2 PubMed Links

Gene Details

Gene
C5AR2
Gene Name
complement component 5a receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271749.1 245 Missense Mutation CAC,CGC H35R NP_001258678.1
NM_001271750.1 245 Missense Mutation CAC,CGC H35R NP_001258679.1
NM_018485.2 245 Missense Mutation CAC,CGC H35R NP_060955.1
XM_011526736.2 245 Missense Mutation CAC,CGC H35R XP_011525038.1
Gene
DHX34
Gene Name
DEAH-box helicase 34
There are no transcripts associated with this gene.

View Full Product Details