Product Details

SNP ID
rs139213903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9186042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGATGCTGGTGAACATCCAGAC[C/T]GAGAACAAAGCCATCTCCTACATGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR7D2 PubMed Links

Gene Details

Gene
OR7D2
Gene Name
olfactory receptor family 7 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175883.2 449 Silent Mutation ACC,ACT T87T NP_787079.1

View Full Product Details