Product Details

SNP ID
rs139270509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15053550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCACCAAAGCCCGGGAAGGTTC[C/T]GAAGAAGACCTGGATGCTCTGGAAC
Phenotype
MIM: 605848
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASP14 PubMed Links

Gene Details

Gene
CASP14
Gene Name
caspase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012114.2 308 Silent Mutation TCC,TCT S32S NP_036246.1
XM_011527861.1 308 Silent Mutation TCC,TCT S32S XP_011526163.1

View Full Product Details