Product Details
- SNP ID
-
rs139322374
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:45669565 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCAGCTGCTGCTGCGGCTCTCACT[G/T]TGCGGGCTGCTGCTCCAGAGGGCGG
- Phenotype
-
MIM: 137241
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GIPR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5390] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GIPR
- Gene Name
- gastric inhibitory polypeptide receptor
- Gene
- MIR642A
- Gene Name
- microRNA 642a
There are no transcripts associated with this gene.
- Gene
- MIR642B
- Gene Name
- microRNA 642b
There are no transcripts associated with this gene.
View Full Product Details