Product Details

SNP ID
rs139533300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9413811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGCCTCTCTCCACTGTGAGTTC[A/G]TGCATGCTTAGTAAGGTCTGAGTTC
Phenotype
MIM: 604751
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF266 PubMed Links

Gene Details

Gene
ZNF266
Gene Name
zinc finger protein 266
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271314.1 1973 Nonsense Mutation CGA,TGA R372* NP_001258243.1
NM_006631.3 1973 Nonsense Mutation CGA,TGA R372* NP_006622.2
XM_005259716.3 1973 Nonsense Mutation CGA,TGA R372* XP_005259773.1
XM_006722620.2 1973 Nonsense Mutation CGA,TGA R439* XP_006722683.1
XM_006722621.2 1973 Nonsense Mutation CGA,TGA R439* XP_006722684.1
XM_006722622.2 1973 Nonsense Mutation CGA,TGA R439* XP_006722685.1
XM_006722623.3 1973 Nonsense Mutation CGA,TGA R439* XP_006722686.1
XM_006722627.2 1973 Nonsense Mutation CGA,TGA R372* XP_006722690.1
XM_006722630.2 1973 Nonsense Mutation CGA,TGA R372* XP_006722693.1
XM_011527644.1 1973 Nonsense Mutation CGA,TGA R439* XP_011525946.1
XM_011527647.1 1973 Nonsense Mutation CGA,TGA R372* XP_011525949.1
XM_011527648.1 1973 Nonsense Mutation CGA,TGA R372* XP_011525950.1
XM_011527650.1 1973 Nonsense Mutation CGA,TGA R372* XP_011525952.1
XM_017026154.1 1973 Nonsense Mutation CGA,TGA R439* XP_016881643.1
XM_017026155.1 1973 Nonsense Mutation CGA,TGA R439* XP_016881644.1
XM_017026156.1 1973 Nonsense Mutation CGA,TGA R372* XP_016881645.1
XM_017026157.1 1973 Nonsense Mutation CGA,TGA R372* XP_016881646.1
XM_017026158.1 1973 Nonsense Mutation CGA,TGA R372* XP_016881647.1
XM_017026159.1 1973 Nonsense Mutation CGA,TGA R372* XP_016881648.1
XM_017026160.1 1973 Nonsense Mutation CGA,TGA R372* XP_016881649.1
XM_017026161.1 1973 Nonsense Mutation CGA,TGA R372* XP_016881650.1
XM_017026162.1 1973 Nonsense Mutation CGA,TGA R372* XP_016881651.1
XM_017026163.1 1973 Nonsense Mutation CGA,TGA R372* XP_016881652.1
XM_017026164.1 1973 Nonsense Mutation CGA,TGA R372* XP_016881653.1
XM_017026165.1 1973 Nonsense Mutation CGA,TGA R372* XP_016881654.1
XM_017026166.1 1973 Nonsense Mutation CGA,TGA R372* XP_016881655.1
XM_017026167.1 1973 Nonsense Mutation CGA,TGA R372* XP_016881656.1
XM_017026168.1 1973 Nonsense Mutation CGA,TGA R142* XP_016881657.1
XM_017026169.1 1973 Nonsense Mutation CGA,TGA R142* XP_016881658.1
XM_017026170.1 1973 Nonsense Mutation CGA,TGA R142* XP_016881659.1
XM_017026171.1 1973 Nonsense Mutation CGA,TGA R142* XP_016881660.1
XM_017026172.1 1973 Nonsense Mutation CGA,TGA R142* XP_016881661.1
XM_017026173.1 1973 UTR 3 XP_016881662.1

View Full Product Details