Product Details

SNP ID
rs139582710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40693009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGACTTCTGCAGGACACAGTCT[C/G]TGTCTCCATCAGCTGCAGCCTTCAC
Phenotype
MIM: 615567 MIM: 604018
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COQ8B PubMed Links

Gene Details

Gene
COQ8B
Gene Name
coenzyme Q8B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142555.2 1540 Missense Mutation ACA,AGA T372R NP_001136027.1
NM_024876.3 1540 Missense Mutation ACA,AGA T413R NP_079152.3
Gene
NUMBL
Gene Name
NUMB like, endocytic adaptor protein
There are no transcripts associated with this gene.

View Full Product Details