Product Details

SNP ID
rs139616778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17560444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGCCCTGAAATCAGCTCGAGA[C/T]ATGTGGGCTGATTACATCCTGGTAA
Phenotype
MIM: 609967
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COLGALT1 PubMed Links

Gene Details

Gene
COLGALT1
Gene Name
collagen beta(1-O)galactosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024656.3 226 Silent Mutation GAC,GAT D156D NP_078932.2
XM_005260080.4 226 Silent Mutation GAC,GAT D73D XP_005260137.1
XM_011528297.2 226 Silent Mutation GAC,GAT D63D XP_011526599.1
XM_011528298.2 226 Silent Mutation GAC,GAT D63D XP_011526600.1
XM_011528299.2 226 Intron XP_011526601.1
Gene
FAM129C
Gene Name
family with sequence similarity 129 member C
There are no transcripts associated with this gene.

View Full Product Details