Product Details

SNP ID
rs139619574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52155469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAGTATGCCTTCTCTGATGGTA[C/T]GTCAGGCCTGTTATATGACTAAAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF836 PubMed Links

Gene Details

Gene
ZNF836
Gene Name
zinc finger protein 836
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102657.1 2623 Silent Mutation ACA,ACG T738T NP_001096127.1
XM_011526558.2 2623 Silent Mutation ACA,ACG T738T XP_011524860.1
XM_011526559.2 2623 Silent Mutation ACA,ACG T738T XP_011524861.1
XM_017026389.1 2623 Intron XP_016881878.1

View Full Product Details