Product Details

SNP ID
rs139651162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:897770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCTCGGGCCCATCTCACTCCC[C/T]GCTGGCACTCATTCCACTGTGCGGA
Phenotype
MIM: 604062
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED16 PubMed Links

Gene Details

Gene
MED16
Gene Name
mediator complex subunit 16
There are no transcripts associated with this gene.

Gene
R3HDM4
Gene Name
R3H domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138774.3 Intron NP_620129.2
XM_011528416.2 Intron XP_011526718.1
Gene
RNU6-9
Gene Name
RNA, U6 small nuclear 9
There are no transcripts associated with this gene.

View Full Product Details