Product Details

SNP ID
rs139695647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2851493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATGAAACTCAATTTAAGGCCAG[C/T]GGGTCAGTTTCTCAGCAGGATATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF555 PubMed Links

Gene Details

Gene
ZNF555
Gene Name
zinc finger protein 555
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172775.1 297 Silent Mutation AGC,AGT S52S NP_001166246.1
NM_152791.4 297 Silent Mutation AGC,AGT S52S NP_690004.4
XM_011527716.2 297 Silent Mutation AGC,AGT S54S XP_011526018.1
XM_017026375.1 297 Silent Mutation AGC,AGT S54S XP_016881864.1

View Full Product Details