Product Details

SNP ID
rs139702020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41425802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTCAAAGGGGAAGGGTGCCACA[C/T]GGGCTGCCGCCCGCAGCAGCCAGGG
Phenotype
MIM: 615357 MIM: 608348
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5SL PubMed Links

Gene Details

Gene
ATP5SL
Gene Name
ATP5S like
There are no transcripts associated with this gene.

Gene
B3GNT8
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198540.2 2750 Missense Mutation CAT,CGT H326R NP_940942.1
XM_011526934.2 2750 Missense Mutation CAT,CGT H326R XP_011525236.1
Gene
BCKDHA
Gene Name
branched chain keto acid dehydrogenase E1, alpha polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000709.3 2750 Intron NP_000700.1
NM_001164783.1 2750 Intron NP_001158255.1

View Full Product Details